missing translation for 'onlineSavingsMsg'
Learn More

MEKK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18396146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396146 100 μL 100µL
18313067 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396146 Supplier Novus Biologicals Supplier No. NBP302962100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEKK1 Polyclonal specifically detects MEKK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Map3k1, MAPK/ERK kinase kinase 1, MAPKKK1MAP/ERK kinase kinase 1, MEK kinase 1, MEKK1EC 2.7.11.25, MEKKMEKK 1, mitogen-activated protein kinase kinase kinase 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1400 to the C-terminus of human MEKK1 (NP_005912.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Breast Cancer, Cancer, Lipid and Metabolism, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 4214
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.