missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEKK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | MEKK1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227343
|
Novus Biologicals
NBP2-58101 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691198
|
Novus Biologicals
NBP2-58101-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEKK1 Polyclonal specifically detects MEKK1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| MEKK1 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Breast Cancer, Cancer, Lipid and Metabolism, MAP Kinase Signaling, mTOR Pathway, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| Map3k1, MAPK/ERK kinase kinase 1, MAPKKK1MAP/ERK kinase kinase 1, MEK kinase 1, MEKK1EC 2.7.11.25, MEKKMEKK 1, mitogen-activated protein kinase kinase kinase 1 | |
| MAP3K1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 4214 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SSSKCDDSFGCSSNSSNAVIPSDETVFTPVEEKCRLDVNTELNSSIEDLLEASMPSSDTTVTFKSEVAVLSPEKAENDDTYKDDVNHNQKCKEKMEAEEEEALAIAMAMSASQDALPIVPQLQVENGEDIIIIQQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title