missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK5, p Ser311, p Thr315 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61105
This item is not returnable.
View return policy
Description
MEK5 Polyclonal specifically detects MEK5 in Rat samples. It is validated for Western Blot.
Specifications
| MEK5 (p Ser311, p Thr315) | |
| Polyclonal | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| HsT17454, MAP kinase kinase 5, MAP kinase kinase MEK5b, MAPK/ERK kinase 5, MAPKK 5, MAPKK5EC 2.7.12.2, MEK5MEK 5, mitogen-activated protein kinase kinase 5, MKK5, PRKMK5dual specificity mitogen-activated protein kinase kinase 5 | |
| Rabbit | |
| 49 kDa | |
| 0.1 mL | |
| Phospho Specific | |
| 5607 | |
| Rat | |
| IgG |
| Western Blot | |
| 0.35 mg/mL | |
| Western Blot 1:1000 | |
| Q62862 | |
| MAP2K5 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser311/Thr315 conjugated to KLH. Accession # Q62862 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the approx. 49k MEK5 protein phosphorylated at Ser311, Thr314. Immunolabeling of the MEK5 band is blocked by preadsorption with the phospho-peptide used as antigen but not be the corresponding dephospho-peptide. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction