missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEK3/MEK6 Recombinant Rabbit Monoclonal Antibody (24GB310)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557061
This item is not returnable.
View return policy
Description
MEK3/MEK6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
Specifications
| MEK3/MEK6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MAP2K3 | |
| AW212142; dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; Map2k3; Map2k6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 3; mitogen-activated protein kinase kinase 6; MKK3; MKK6; mMKK3b; OTTHUMP00000166044; Prkmk3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK2; SKK3; Stress-activated protein kinase kinase 2; stress-activated protein kinase kinase 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114495, 26397, 26399, 303200, 5606, 5608 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB310 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09110, P46734, P52564, P70236 | |
| MAP2K3, MAP2K6 | |
| A synthesized peptide derived from human MEK3/MEK6 (aa 300-347). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction