missing translation for 'onlineSavingsMsg'
Learn More

MEK2 Rabbit anti-Human, Mouse, Clone: 6D5K1, Novus Biologicals™

Product Code. 18349923 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349923 20 μg 20µL
18312257 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18349923 Supplier Novus Biologicals Supplier No. NBP31565120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MEK2 Monoclonal antibody specifically detects MEK2 in Human, Mouse samples. It is validated for Western Blot, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK2
Applications Western Blot, KnockDown
Classification Monoclonal
Clone 6D5K1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEK2 (P36507).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Kinase, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5605
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.