missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK2 Antibody (OTI8G6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72646PCP
This item is not returnable.
View return policy
Description
MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| MEK2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| MAP2K2 | |
| Full-length protein expressed in 293T cell transfected with human MEK2 expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI8G6 | |
| Flow Cytometry | |
| ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction | |
| 5605.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction