missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK1 Antibody (OTI1F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47833
This item is not returnable.
View return policy
Description
MEK1 Monoclonal specifically detects MEK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MEK1 | |
| Monoclonal | |
| 0.99 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| MAP2K1 | |
| Full-length protein expressed in 293T cell transfected with human MAP2K1 expression vector | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens mitogen-activated protein kinase kinase 1 (MAP2K1). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1F5 | |
| Unconjugated | |
| dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
| Mouse | |
| 43.4 kDa | |
| 0.1 mL | |
| Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
| 5604 | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction