missing translation for 'onlineSavingsMsg'
Learn More

MEK1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18417996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417996 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417996 Supplier Novus Biologicals Supplier No. NBP224560AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEK1 Polyclonal antibody specifically detects MEK1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human MEK1 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.