missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Meisoindigo

Product Code. 17332501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17332501 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17332501 Supplier Thermo Scientific™ Supplier No. 468801000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

Chemical Name or Material Meisoindigo
CAS 97207-47-1
Synonym N-Methylisoindigotin; 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one
Quantity 100 mg
Melting Point 230°C
Molecular Formula C17 H12 N2 O2
Formula Weight 276.29
Physical Form Crystalline Powder
Color Orange to Dark-red to Brown
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.