missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEIS3 Monoclonal Antibody (4E12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524479
This item is not returnable.
View return policy
Description
Peptide Sequence: MARRYDELPH YPGIVDGPAA LASFPETVPA VPGPYGPHRP PQPLPPGLDS DGLKREKDEI YGHPLFPLLA LVFEKCELAT CSPRDGAGAG LGTPPGGDVC SSDSFNEDIA.
Transcriptional regulator which directly modulates PDPK1 expression, thus promoting survival of pancreatic beta-cells. Also regulates expression of NDFIP1, BNIP3, and CCNG1.
Specifications
| MEIS3 | |
| Monoclonal | |
| Unconjugated | |
| MEIS3 | |
| AI573393; Homeobox protein Meis3; Meis homeobox 3; Meis homeobox 3 pseudogene 1; Meis1, myeloid ecotropic viral integration site 1 homolog 3; meis1-related protein 2; Meis3; MEIS3P1; Mrg2; myeloid ecotropic viral integration site-related gene 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 56917 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4E12 | |
| PBS with no preservative; pH 7.4 | |
| Q99687 | |
| MEIS3 | |
| MEIS3 (NP_001009813.1, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction