missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEIS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112606
This item is not returnable.
View return policy
Description
MEIS2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.
Specifications
| MEIS2 | |
| Polyclonal | |
| Unconjugated | |
| MEIS2 | |
| A430109D20Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; homeobox protein Meis2; HsT18361; Meis homeobox 2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; meis1-related protein 1; MEIS2; Meis2 homeobox; MGC2820; MRG1; myeloid ecotropic viral integration site-related gene 1; OTTHUMP00000160209; Stra10; TALE homeobox protein Meis2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 17536, 4212 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.68 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O14770, P97367 | |
| MEIS2 | |
| Recombinant Human Homeobox protein Meis2 protein (377-470AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction