missing translation for 'onlineSavingsMsg'
Learn More

MEIS1 Antibody (OTI3H2), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

£195.00 - £417.00

Specifications

Antigen MEIS1
Clone OTI3H2
Dilution Western Blot 1:2000, Immunohistochemistry 1:500
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30306212
View Documents
Novus Biologicals
NBP3-44994-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30306327
View Documents
Novus Biologicals
NBP3-44994-30ul
30 μL
£195.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

MEIS1
Western Blot 1:2000, Immunohistochemistry 1:500
Monoclonal
Purified
RUO
PBS (pH 7.3) 1% BSA, 50% glycerol
4211
IgG1
>95%
Antibody
OTI3H2
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse)
Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1(NP_002389) produced in E.coli.
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.