missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEIOB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568422
This item is not returnable.
View return policy
Description
MEIOB Polyclonal Antibody for IHC (P)
Single-stranded DNA-binding protein required for homologous recombination in meiosis I. Required for double strand breaks (DSBs) repair and crossover formation and promotion of faithful and complete synapsis. Not required for the initial loading of recombinases but required to maintain a proper number of RAD51 and DMC1 foci after the zygotene stage. May act by ensuring the stabilization of recombinases, which is required for successful homology search and meiotic recombination. Displays Single-stranded DNA 3'-5' exonuclease activity in vitro.
Specifications
| MEIOB | |
| Polyclonal | |
| Unconjugated | |
| MEIOB | |
| 4930528F23Rik; C16orf73; C6H16orf73; gs129; Meiob; meiosis specific with OB domains; meiosis specific with OB-fold; meiosis-specific with OB domain-containing protein; MLZ-675; Protein expressed in male leptotene and zygotene spermatocytes 675 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 254528 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8N635 | |
| MEIOB | |
| Synthetic protein corresponding to residues near the C terminal of human MEIOB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction