missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEIG1 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-21156 can be used with blocking peptide PEP-1270.
MEIG1, a murine gene first identified as a testis specific gene, is a chromosome/chromatin-binding protein initially expressed during meiosis but retained in the germ cell nucleus throughout later stages of spermatogenesis. MEIG1 is a highly conserved basal metazoan gene that is indispensable for mouse spermatogenesis. It is important for normal meiotic differentiation and absolutely crucial for terminal differentiation of spermatozoa. MEIG1 encodes two alternative transcripts, designated 2a2 and 11a2, both of which encode for a common ORF but differing in their 5' untranslated region (5'UTR) due to alternative promoters.
Specifications
Specifications
| Antigen | MEIG1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MEIG1 |
| Gene Accession No. | Q5JSS6, Q61845 |
| Gene Alias | bA2K17.3; Meg1; Meig1; meiosis expressed gene 1; meiosis expressed gene 1 homolog; meiosis expressed gene 1 protein homolog; meiosis/spermiogenesis associated 1; Meiosis-expressed gene 1 protein; MLZ-278; Protein expressed in male leptotene and zygotene spermatocytes 278; SPATA39; spermatogenesis associated 39 |
| Gene Symbols | MEIG1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?