missing translation for 'onlineSavingsMsg'
Learn More

MEIG1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18335931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335931 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335931 Supplier Novus Biologicals Supplier No. NBP281733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEIG1 Polyclonal specifically detects MEIG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIG1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias bA2K17.3, meiosis expressed gene 1 homolog, meiosis expressed gene 1 homolog (mouse)
Gene Symbols MEIG1
Host Species Rabbit
Immunogen MEIG1 antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminal of human MEIG1. The immunogen is located within the last 50 amino acids of MEIG1. Amino Acid Squence: YVKKLQRRDNTFYYYNKQRE
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.