missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ MEGM™ Mammary Epithelial Cell Growth Medium SingleQuots™ Kit

Product Code. 11605500
Change view
Click to view available options
BPE: 2.0 mL, hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11605500 BPE: 2.0 mL, hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11605500 Supplier Lonza™ Supplier No. CC4136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplements and growth factors kit for use with mammary epithelial cell basal medium.

  • Separately sold SingleQuot™ kit for supplementing basal medium.
  • Part of the full MEGM™ Mammary Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3150), which consists of basal medium and supplements packaged together.
  • Optimized for the growth of mammary epithelial cells in a serum-free environment.
  • This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
  • Includes the following:
    • 1 x Orange Cap Vial with BPE 2.00 mL
    • 1 x Green Cap Vial with hEGF 0.50 mL
    • 1 x Lilac Cap Vial with Insulin 0.50 mL
    • 1 x Natural Cap Vial with Hydrocortisone 0.50 mL
    • 1 x Red Cap Vial with GA-1000 0.50 mL
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Product Type Supplements and growth factors
Quantity BPE: 2.0 mL, hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL
Content And Storage BPE, hEGF, Insulin, Hydrocortisone, GA-1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.