missing translation for 'onlineSavingsMsg'
Learn More

MEGF9 Antibody [PerCP], Novus Biologicals™

Product Code. 30736858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736858 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736858 Supplier Novus Biologicals Supplier No. AF7768C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

MEGF9 Polyclonal antibody specifically detects MEGF9 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEGF9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias EGFL5KIAA0818, EGF-like protein 5, EGF-like-domain, multiple 5, Epidermal growth factor-like protein 5, Multiple EGF-like domains protein 9, multiple EGF-like-domains 9, multiple epidermal growth factor-like domains protein 9
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human MEGF9, Ala36-Asn514, Accession # Q9H1U4
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1955
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.