missing translation for 'onlineSavingsMsg'
Learn More

MEGF9 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30736908 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736908 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736908 Supplier Novus Biologicals Supplier No. AF7768D

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Sheep Polyclonal Antibody

MEGF9 Polyclonal antibody specifically detects MEGF9 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen MEGF9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias EGFL5KIAA0818, EGF-like protein 5, EGF-like-domain, multiple 5, Epidermal growth factor-like protein 5, Multiple EGF-like domains protein 9, multiple EGF-like-domains 9, multiple epidermal growth factor-like domains protein 9
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human MEGF9, Ala36-Asn514, Accession # Q9H1U4
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1955
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.