missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MEGF8 Polyclonal specifically detects MEGF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MEGF8 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | C19orf49, chromosome 19 open reading frame 49, EGFL4MGC120684, EGF-like domain-containing protein 4, EGF-like protein 4, EGF-like-domain, multiple 4, Epidermal growth factor-like protein 4, FLJ22365, HBV pre-s2 binding protein 1, KIAA0817, MGC138147, multiple EGF-like domain protein 4, Multiple EGF-like domains protein 8, multiple EGF-like-domains 8, multiple epidermal growth factor-like domains 8, multiple epidermal growth factor-like domains protein 8, SBP1 |
| Gene Symbols | MEGF8 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the amino acids: PCSPMPRSPEECRRLRTCSECLARHPRTLQPGDGEASTPRCKWCTNCPEGACIGRNGSCTSENDCRINQREVFWAGNCSEAACGAADCEQCTREGKCMWTRQFKRTGETRRILSVQPTYDWTCFSHSLLNVS |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?