missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEGF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103934
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MEGF6.
MEGF6 gene ontology annotations related to this gene include calcium ion binding.
Specifications
| MEGF6 | |
| Polyclonal | |
| Unconjugated | |
| MEGF6 | |
| 2600001P17Rik; Egfl3; EGF-like domain-containing protein 3; EGF-like protein 3; EGF-like-domain, multiple 3; Epidermal growth factor-like protein 3; KIAA0815; MEGF6; multiple EGF like domains 6; multiple EGF-like domain protein 3; multiple EGF-like domains protein 6; multiple EGF-like-domains 6; multiple epidermal growth factor-like domains 6; multiple epidermal growth factor-like domains protein 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1953, 230971, 65049 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75095, O88281, Q80V70 | |
| MEGF6 | |
| A synthesized peptide derived from human MEGF6(Accession O75095), corresponding to amino acid residues P141-F191. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction