missing translation for 'onlineSavingsMsg'
Learn More

Megestrol Acetate, TRC

Product Code. 30137836
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137836 1 g 1g
30003423 100 mg 100mg
30012329 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137836 Supplier Toronto Research Chemicals Supplier No. TRCM2080501G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-33-5
Molecular Formula C24 H32 O4
Molecular Weight (g/mol) 384.51
Synonym Megestrol Acetate, 6-Methyl-3,20-dioxopregna-4,6-dien-17-yl Acetate, Medroxyprogesterone Acetate Imp. G (EP), 17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 17-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 17α-Acetoxy-6-dehydro-6-methylprogesterone, 17α-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17α-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 5071, 6-Dehydro-6-methyl-17α-acetoxyprogesterone, 6-Methyl-17α-acetoxy-4,6-pregnadiene-3,20-dione, 6-Methyl-17α-hydroxy-δ6-progesterone acetate, 6-Methyl-6-dehydro-17α-acetoxyprogesterone, 6-Methyl-δ4,6-pregnadien-17α-ol-3,20-dione acetate, BDH 1298, DMAP, MGA, Magestin, Maygace, Megace, Megalia, Megeron, Megestat, Megestil, Megestin, Megestrol acetate, Megestryl acetate, NSC 71423, Nia, Niagestin, Ovaban, Ovarid, SC 10363
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1
Formula Weight 384.2301
Percent Purity >95%
Chemical Name or Material Megestrol-17-acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.