missing translation for 'onlineSavingsMsg'
Learn More

Megestrol Acetate, MP Biomedicals™

Product Code. 11401701 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401701 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401701 Supplier MP Biomedicals Supplier No. 0219023501
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 595-33-5
Molecular Formula C24H32O4
Molecular Weight (g/mol) 384.516
InChI Key RQZAXGRLVPAYTJ-GQFGMJRRSA-N
Synonym megestrol acetate, niagestin, megace es, megestryl acetate, megace, megeron, ovaban, ovarid, pallace, magestin
PubChem CID 11683
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1=CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

Quantity 1 g
Formula Weight 384.5
Physical Form Solid
Chemical Name or Material Megestrol Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.