Learn More
Invitrogen™ MegaScript™ T7 Transcription Kit Plus
Descrizione
The MegaScript T7 Transcription Kit Plus contains all the reagents and buffers necessary for in vitro synthesis of large amounts of RNA. Each 20-μL reaction yields an average of 100 μg of RNA, while the scaled-up protocol yields up to 1 mg of RNA. This kit is suitable for full- or partially-modified-nucleotide substitution to produce modified RNA. It is also suitable for co-transcriptional capping with cap analogs to make capped RNA that can be translated into protein. Easily scale from RUO to GMP with the incorporation of TheraPure reagents, eliminating reoptimization during scale up.
Features of the MegaScript T7 Transcription Kit Plus include:
- Optimized to achieve high mRNA yields (>5 mg/mL)
- Flexible—allows use of modified nucleotides and/or cap analogs such as ARCA or CleanCap
- Includes GFP control template
- Made up of high quality reagents (TheraPure); eliminates the need to qualify new reagents during scale-up
- Available in two sizes: 50 and 1000 reactions
The MegaScript T7 Transcription Kit Plus requires a linearized DNA template with a T7 RNA polymerase promoter to create >5 mg/mL of RNA. MEGAscript kits are intended for the synthesis of large amounts of unlabeled or low-specific-activity RNA for a variety of uses, including transfections, preclinical mRNA therapeutic mRNA studies, microinjections, in vitro translation, and RNA structure and function analysis. Note that this kit does not create capped RNA, which is necessary for translational studies. MEGAscript kits are not recommended for synthesis of high-specific-activity probes.
Specifica
Specifica
| Cell Type | HEK293, 293, HeLa, CHO-K1, CHO, Jurkat, Primary Human T Cells, Primary Cells, Stem Cells, iPSC, NIH-3T3, HepG2, A549, HUVEC |
| Final Product Type | Synthetic mRNA |
| Format | Animal Origin Free Kit |
| Isolation Technology | Silica Spin Column |
| Purification Target | mRNA |
| Sample Type | mRNA, Total RNA |
| No. of Reactions | 25 Reactions |
| Product Line | MEGAscript |
| Product Type | T7 In Vitro Transcription Kit |
| Quantity | 25 Reactions |
| Vedi altri risultati |
For Research Use Only.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.