missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Megaplex™ RT Primers, Rodent Pool Set v3.0
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 reactions
Unit Size:
50 reactions
Description
Designed to streamline experimental workflows, Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysis on a TaqMan™ Array MicroRNA Card. The Rodent Pool Set v3.0 contains two Megaplex™ RT Primer Pools, Pools A and B. Collectively the pools cover 641 and 373 unique microRNAs for mouse and rat respectively. Each pool also contains 4 species specific controls. The microRNA targets represented in Rodent Pool A tend be better characterized, more broadly expressed and⁄or expressed at a higher level when compared to the Rodent Pool B while the microRNA targets represented in Rodent Pool B tend to be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to the Rodent Pool A. Rodent Pools A and B are intended for use with the TaqMan™ Rodent MicroRNA Array Set v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Primer | RT Primers |
| Product Line | Megaplex, TaqMan |
| Product Type | RT Primer |
| Quantity | 50 reactions |
| Species | Rat, Mouse |
| Sufficient For | 50 Reactions |
| For Use With (Application) | miRNA analysis |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction