missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Megaplex™ Primer Pools, Rodent Pools Set v3.0

Product Code. 12067789 Shop All Applied Biosystems Products
Click to view available options
Quantity:
50 reactions
Unit Size:
50 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12067789

Brand: Applied Biosystems™ 4444766

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Megaplex™ Primer Pools, Rodent Pools Set v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools Sets.

Species: Mouse, Rat
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C
GC-Rich PCR Performance Low
PCR Method qPCR
Primer RT Primers, PreAmp Primers
Product Line Megaplex, TaqMan
Product Type Primer Pool
Quantity 50 reactions
Species Rat, Mouse
Sufficient For 50 Reactions
For Use With (Application) miRNA analysis
No. of Reactions 50 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.