missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Megaplex™ Primer Pools, Rodent Pools A
GREENER_CHOICE

Product Code. 10724407 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10724407 50 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10724407 Supplier Applied Biosystems™ Supplier No. 4401090
  • ONLINE EXCLUSIVE PRICE
    £966.00
    £743.82 / 1 sets
    Save £222.18
    23% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Megaplex™ Primer Pools, Rodent Pools A

Applied Biosystems™ Megaplex™ Primer Pools, Rodent Pool A is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools A. Megaplex™ RT Primers, Rodent Pools A contains RT primers for 335 and 226 unique microRNAs for mouse and rat respectively plus 4 species specific controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Rodent Pool B. Rodent Pool A is intended for use with the TaqMan™ Rodent MicroRNA A Array v2.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage Contains 150μL of 10X primers (Megaplex™ PreAmp Primers, Rodent Pool A) and 50μL of 10X primer (Megaplex™ RT Primers, Rodent Pool A). Store at -15 to -25°C.
GC-Rich PCR Performance High
PCR Method qPCR
Primer RT Primers, PreAmp Primers
Product Line Megaplex, TaqMan
Product Type Primer Pool
Quantity 50 reactions
Species Rat, Mouse
Sufficient For 50 Reactions
For Use With (Application) miRNA analysis
Fidelity (vs. Taq) 10 X
No. of Reactions 50 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.