missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Megaplex™ Primer Pools, Human Pools A v2.1
GREENER_CHOICE

Product Code. 10585575 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
50 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10585575 50 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10585575 Supplier Applied Biosystems™ Supplier No. 4401009
  • ONLINE EXCLUSIVE PRICE
    £998.00
    £768.46 / Pack of 1
    Save £229.54
    23% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Megaplex™ Primer Pools, Human Pools A v2.1

Megaplex™ Primer Pools, Human Pools A v2.1 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex PreAmp Primers, Human Pools A v2.1. Megaplex™ RT Primers, Human Pool A v2.1 contains RT primers for 377 unique microRNAs and 4 controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Human Pool B. Human Pool A is intended for use with the TaqMan™ Array Human MicroRNA A Card v2.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
GC-Rich PCR Performance High
PCR Method qPCR
Primer RT Primers, PreAmp Primers
Product Line Megaplex, TaqMan
Product Type Primer Pool
Quantity 50 reactions
Species Human
Sufficient For 50 Reactions
For Use With (Application) miRNA analysis
No. of Reactions 50 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.