missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Megaplex™ PreAmp Primers, Human Pool B v3.0
GREENER_CHOICE

Product Code. 11650089 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 reactions
Unit Size:
50 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11650089

Brand: Applied Biosystems™ 4444303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Megaplex™ PreAmp Primers, Human Pool B v3.0

For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. With content matched to the Megaplex™ RT Primers, Megaplex™ PreAmp Primers, Human Pool B v3.0 is designed to be used with Megaplex™ RT Primers, Human Pool B v3.0. Following reverse transcription using Megaplex™ RT Primers, Megaplex™ PreAmp Primers along with the TaqMan™ PreAmp Master Mix uniformly amplify all microRNAs prior to quantitation by real-time PCR using TaqMan™ Array MicroRNA Cards.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C
GC-Rich PCR Performance Low
PCR Method qPCR
Green Features Sustainable packaging
Primer PreAmp Primers
Product Line Megaplex
Product Type Preamp Primer
Quantity 50 reactions
Shipping Condition Room Temperature
Species Human
Sufficient For 50 Reactions
No. of Reactions 50 reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.