missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MegaCult™-C Medium Without Cytokines

Product Code. 17108241
Encompass
Click to view available options
Quantity:
24 x 1.7 mL
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 17108241

Brand: STEMCELL Technologies 04900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium without cytokines

  • MegaCult™-C Complete Kit Without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining
  • The addition of appropriate cytokines, as chosen by the user, is required

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 24 x 1.7 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.