missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MegaCult™-C Human Serum

Product Code. 17178231
Encompass
Change view
Click to view available options
Quantity:
6 mL
Unit Size:
6mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17178231 6 mL 6mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17178231 Supplier STEMCELL Technologies Supplier No. 04807

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Human serum for dilutions

  • MegaCult™-C Staining Kit for CFU-Mk contains all the reagents needed to stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium
  • Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system
  • The kit is also suitable for detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
Quantity 6 mL
For Use With (Application) Colony Assay; Immunocytochemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.