missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MegaCult™-C Complete Kit Without Cytokines
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MegaCult™-C Complete Kit Without Cytokines includes all reagents (except cytokines) required for colony-forming unit (CFU) assays of megakaryocyte progenitor cells (CFU-Mk) in a collagen-based culture medium, including the detection of human CFU-Mk colonies by immunocytochemical staining
- The addition of appropriate cytokines, as chosen by the user, is required
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction