missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MegaCult™-C Collagen and Medium Without Cytokines
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MegaCult™-C Collagen and Medium Without Cytokines kit contains medium and collagen solution for colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk)
- The addition of cytokines is required
- MegaCult™-C medium is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples, after addition of appropriate cytokines
- It is suitable for use with CD34+ enriched cells, mononuclear cells, and cells isolated by other purification methods
- It is also intended for assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow, after addition of appropriate cytokines
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction