missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MegaCult™-C Collagen and Medium with Lipids
Description
- MegaCult™-C Collagen and Medium with Lipids kit includes medium and collagen solution for colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk)
- The addition of cytokines is required
- MegaCult™-C Medium with Lipids is intended for the culture of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples after addition of appropriate cytokines
- It is recommended for use with CD34+-enriched cells, mononuclear cells, and cells isolated by other purification methods
- It can also be used for CFU-MK assays of megakaryocyte progenitor cells in unseparated or purified cell suspensions of mouse bone marrow after addition of appropriate cytokines
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?