missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MegaCult™-C Collagen and Medium with Cytokines
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- MegaCult™-C Collagen and Medium with Cytokines kit contains medium and collagen solution for colony-forming unit (CFU) assays of human megakaryocyte progenitor cells (CFU-Mk)
- The medium contains recombinant human (rh) IL-3, IL-6, and thrombopoietin
- MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood, and cord blood samples
- It is suitable for use with CD34+ enriched cells, mononuclear cells, and cells isolated by other purification methods
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction