missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583579
This item is not returnable.
View return policy
Description
Immunogen sequence: LATWNFQFLP QVNGHPDPFD EIILFSLPPA LDWDSVHVPG PHAMTIQELV DYVNARQKQG IYEEYEDIRR ENPVGTFHCS MSPGNL Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 97%.
MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.
Specifications
| MEG2 | |
| Polyclonal | |
| Unconjugated | |
| PTPN9 | |
| Meg2; meg-2; protein tyrosine phosphatase, non-receptor type 9; protein-tyrosine phosphatase MEG2; Protein-tyrosine phosphatase, non-receptor type 9 (Protein-tyrosine phosphatase MEG2) (PTPase-MEG2); PTPase MEG2; PTPase-MEG2; PTPMEG2; PTPN9; Tyrosine-protein phosphatase non-receptor type 9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43378 | |
| PTPN9 | |
| Recombinant protein corresponding to Human PTPN9. Recombinant protein control fragment (Product #RP-98128). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction