missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEFV Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-80113AF350
This item is not returnable.
View return policy
Description
MEFV Polyclonal antibody specifically detects MEFV in Human samples. It is validated for Western Blot
Specifications
| MEFV | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| FMF, FMFmarenostrin, Marenostrin, Mediterranean fever, MEFpyrin, MGC126560, MGC126586, TRIM20 | |
| Synthetic peptide corresponding to aa 2-29 (A2KTPSDHLLSTLEELVPYDFEKFKFKLQ29) of human MEFV. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction | |
| 4210 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction