missing translation for 'onlineSavingsMsg'
Learn More

Mefloquine hydrochloride

Product Code. 15668715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15668715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15668715 Supplier Thermo Scientific Acros Supplier No. 461130010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 51773-92-3 | C17H17ClF6N2O | 414.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 51773-92-3
Melting Point 253.0°C
Infrared Spectrum Authentic
Molecular Formula C17H17ClF6N2O
MDL Number MFCD00797519,MFCD00797519
Quantity 1 g
Synonym mefloquine hcl, r-2,8-bis trifluoromethyl quinolin-4-yl s-piperidin-2-yl methanol hydrochloride, r*,s*-2,8-bis-trifluoromethyl-quinolin-4-yl-piperidin-2-yl-methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl-2s-piperidin-2-yl methanol;hydrochloride, as-rel-a-2r-2-piperidinyl-2,8-bis trifluoromethyl-4-quinolinemethanol monohydrochloride, +-mefloquine hydrochloride, unii-5y9l3636o3 component, r-2,8-bis trifluoromethyl-4-quinolinyl 2s-2-piperidinyl methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl 2s-piperidin-2-yl methanol hydrochloride, s-2,8-bis trifluoromethyl quinolin-4-yl r-piperidin-2-yl methanolhydrochloride
InChI Key WESWYMRNZNDGBX-UHFFFAOYNA-N
SMILES [H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride
Molecular Weight (g/mol) 414.78
PubChem CID 18660697
Formula Weight 414.77
Percent Purity ≥97.5% (HPLC) ≥97.5% (on dry substance) (Argentometry)
Chemical Name or Material Mefloquine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Mefloquine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Mefloquine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.