missing translation for 'onlineSavingsMsg'
Learn More

Mefloquine hydrochloride, MedChemExpress

Product Code. 16491856
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16491856

Brand: Medchem Express HY17437A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mefloquine hydrochloride (Mefloquin hydrochloride), a quinoline antimalarial agent, is an anti-SARS-CoV-2 entry inhibitor. Mefloquine hydrochloride is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ∼1 μM. Mefloquine hydrochloride can be used for malaria, systemic lupus erythematosus and cancer research.

Mefloquine hydrochloride (Mefloquin hydrochloride), a quinoline antimalarial agent, is an anti-SARS-CoV-2 entry inhibitor. Mefloquine hydrochloride is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ∼1 μM. Mefloquine hydrochloride can be used for malaria, systemic lupus erythematosus and cancer research.

Specifications

Chemical Name or Material Mefloquine hydrochloride
Physical Form Solid
CAS 51773-92-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C17H17ClF6N2O
For Use With (Application) COVID-19-immunoregulation
Synonym Mefloquin hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (241.10 mM) ∣H2O : 2.86 mg/mL (6.90 mM; ultrasonic and warming and heat to 60°C)
SMILES O[C@H]([C@]1([H])NCCCC1)C2=CC(C(F)(F)F)=NC3=C(C(F)(F)F)C=CC=C32.[H]Cl
Molecular Weight (g/mol) 414.77
Formula Weight 414.77
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.