missing translation for 'onlineSavingsMsg'
Learn More

Mefloquine hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325811
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325811 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325811 Supplier Dr. Ehrenstorfer Supplier No. DREA14860900AL100
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Mefloquine hydrochloride
CAS 51773-92-3
Analyte or Component Names Mefloquine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H16 F6 N2 O . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Mefloquine hydrochloride, (RS)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2SR)-piperidin-2-yl]methanol hydrochloride
InChI Formula InChI=1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15+;/m0./s1
SMILES Cl.O[C@@H]([C@@H]1CCCCN1)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F
IUPAC Name (R)-[2,8-bis(trifluoromethyl)quinolin-4-yl]-[(2S)-piperidin-2-yl]methanol;hydrochloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 414.77
Quantity 1 mL
Formula Weight 414.0934 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.