missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic Acid, Mikromol™

Product Code. 17993976
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17993976

Brand: Mikromol MM0327.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mefenamic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mefenamic acid
CAS 61-68-7
Analyte or Component Names Mefenamic acid
Shipping Condition Room Temperature
Molecular Formula C15 H15 N O2
Certifications/Compliance ISO 17034
Synonym Mefenamic Acid, Anthranilic acid, N-2,3-xylyl- (8CI), 2-[(2,3-Dimethylphenyl)amino]benzoic acid, 2-(2,3-Dimethylanilino)benzoic acid, 2',3'-Dimethyl-N-phenylanthranilic acid, Bafameritin M, Baphameritin M, Bonabol, C.I. 473, CN 35355, Coslan, INF 3355, In-M, Lysalgo, Mefacit, Mefenacid, Mefenamic acid, Mephenamic acid, N-(2,3-Dimethylphenyl)anthranilic acid, N-(2,3-Xylyl)-2-aminobenzoic acid, N-(2,3-Xylyl)anthranilic acid, NSC 94437, Namphen, Parkemed, Ponalar, Ponstan, Ponstel, Ponstil, Ponstyl, Pontal, Relafan, Tanston, Vialidon
InChI Formula InChI=1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)
SMILES Cc1cccc(Nc2ccccc2C(=O)O)c1C
IUPAC Name 2-(2,3-dimethylanilino)benzoic acid
Molecular Weight (g/mol) 241.29
Quantity 250 mg
Formula Weight 241.1103 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.