missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mefenamic Acid, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Mefenamic acid |
| CAS | 61-68-7 |
| Analyte or Component Names | Mefenamic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H15 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Mefenamic Acid, Anthranilic acid, N-2,3-xylyl- (8CI), 2-[(2,3-Dimethylphenyl)amino]benzoic acid, 2-(2,3-Dimethylanilino)benzoic acid, 2',3'-Dimethyl-N-phenylanthranilic acid, Bafameritin M, Baphameritin M, Bonabol, C.I. 473, CN 35355, Coslan, INF 3355, In-M, Lysalgo, Mefacit, Mefenacid, Mefenamic acid, Mephenamic acid, N-(2,3-Dimethylphenyl)anthranilic acid, N-(2,3-Xylyl)-2-aminobenzoic acid, N-(2,3-Xylyl)anthranilic acid, NSC 94437, Namphen, Parkemed, Ponalar, Ponstan, Ponstel, Ponstil, Ponstyl, Pontal, Relafan, Tanston, Vialidon |
| InChI Formula | InChI=1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18) |
| Solution Type | Neat |
| SMILES | Cc1cccc(Nc2ccccc2C(=O)O)c1C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction