Learn More
Mefenamic acid, 98%, Thermo Scientific™
£38.10 - £98.29
Chemical Identifiers
| CAS | 61-68-7 |
|---|---|
| Molecular Formula | C15H15NO2 |
| Molecular Weight (g/mol) | 241.29 |
| MDL Number | MFCD00051721 |
| InChI Key | HYYBABOKPJLUIN-UHFFFAOYSA-N |
| Synonym | mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid |
| PubChem CID | 4044 |
| ChEBI | CHEBI:6717 |
| IUPAC Name | 2-(2,3-dimethylanilino)benzoic acid |
| SMILES | CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15415029
|
Thermo Scientific Alfa Aesar
J62705.14 |
25g |
£38.90 £38.10 / 25g Save £0.80 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15425029
|
Thermo Scientific Alfa Aesar
J62705.22 |
100g |
£103.00 £98.29 / 100g Save £4.71 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.
Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| C15H15NO2 | |
| MFCD00051721 | |
| mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid | |
| CHEBI:6717 | |
| CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C |
Specifications
| 61-68-7 | |
| MFCD00051721 | |
| mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid | |
| HYYBABOKPJLUIN-UHFFFAOYSA-N | |
| 2-(2,3-dimethylanilino)benzoic acid | |
| 4044 | |
| 241.29 | |
| Mefenamic acid |
| C15H15NO2 | |
| 14,5798 | |
| It is soluble in acetone,chloroform,dichloromethane,methanol. Insoluble in water. | |
| CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C | |
| 241.29 | |
| CHEBI:6717 | |
| 98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 200-513-1
missing translation for 'rtecsNumber' : CB4550000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.