missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MEF2D Monoclonal antibody specifically detects MEF2D in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MEF2D |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PCRP-MEF2D-3A4 |
| Conjugate | Biotin |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | DKFZp686I1536, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancerfactor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein, myocyte-specific enhancer factor 2D |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human MEF2D protein (Uniprot: Q14814) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?