missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2C Antibody (CL0369), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-30671-25ul
This item is not returnable.
View return policy
Description
MEF2C Monoclonal specifically detects MEF2C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MEF2C | |
| Monoclonal | |
| Unconjugated | |
| Q06413 | |
| MEF2C | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PPNFEMPVSIPVSSHNSLVYSNPVSSLGNPNLLPLAHPSLQRNSMSPGVTHRPPSAGNTGGLMGGDLTSGAGTSAGNGYGNPRNSPGLLVSPGNLNKNMQAKSPPPMNLGMNNRKPDLRVLIPPGSKNTMPSVNQRINN | |
| 25 μL | |
| Neuroscience | |
| 4208 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL0369 | |
| Western Blot 1 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction