missing translation for 'onlineSavingsMsg'
Learn More

MEF2B Antibody (PCRP-MEF2B-2F9), PerCP, Novus Biologicals™

Product Code. 18361447 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361447 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361447 Supplier Novus Biologicals Supplier No. NBP314207PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MEF2B Monoclonal antibody specifically detects MEF2B in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2B
Applications Flow Cytometry
Classification Monoclonal
Clone PCRP-MEF2B-2F9
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B
Host Species Mouse
Immunogen Recombinant full-length human MEF2B protein (Uniprot: Q02080)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4207
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.