missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2B Antibody (PCRP-MEF2B-2F9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14207CL1
This item is not returnable.
View return policy
Description
MEF2B Monoclonal antibody specifically detects MEF2B in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/Immunofluorescence
Specifications
| MEF2B | |
| Monoclonal | |
| CoraFluor 1 | |
| LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
| Recombinant full-length human MEF2B protein (Uniprot: Q02080) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-MEF2B-2F9 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4207 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction