missing translation for 'onlineSavingsMsg'
Learn More

MEF2A Rabbit anti-Human, Mouse, Rat, Clone: S04-2C8, Novus Biologicals™

Product Code. 18063094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063094 25 μg 25µL
18320892 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063094 Supplier Novus Biologicals Supplier No. NBP31495325UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MEF2A Monoclonal antibody specifically detects MEF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEF2A
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S04-2C8
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1
Host Species Rabbit
Immunogen A synthetic peptide of human MEF2A (Uniprot#: Q02078)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4205
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.