missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEEP Dissolved Gases LCS (methane, ethane, ethene and propane) @ 5 ug/mL in Water, VHG™
Click to view available options
Quantity:
40 mL
Unit Size:
40mL
Description
MEEP Dissolved Gases LCS (methane, ethane, ethene and propane) @ 5 ug/mL in Water
Specifications
Specifications
| Chemical Name or Material | Ethane/Ethene/Methane/N-Propane |
| CAS | 74-84-0, 74-85-1, 74-82-8, 74-98-6 |
| Methods | GFAA |
| Analyte or Component Names | Ethane/Ethene/Methane/N-Propane |
| Shipping Condition | Blue Ice |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Water |
| Quantity | 40 mL |
| Concentration or Composition (by Analyte or Components) | Ethane 5 ppm/Ethene 5 ppm/Methane 5 ppm/N-Propane 5 ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction