missing translation for 'onlineSavingsMsg'
Learn More

Medrysone, MedChemExpress

Product Code. 16408596
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16408596

Brand: Medchem Express HYB1076

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Medrysone is a corticosteroid, in ophthalmology for the treatment of eye inflammations.

Medrysone is a corticosteroid, in ophthalmology for the treatment of eye inflammations.

Specifications

Chemical Name or Material Medrysone
CAS 2668-66-8
Color White
Purity Grade Notes Research
Molecular Formula C22H32O3
Quantity 100 mg
Synonym HMS 6α-Methyl-11β-hydroxyprogesterone
Solubility Information DMSO : 50 mg/mL (145.14 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : ≥ 25 mg/mL (72.57 mM) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES CC([C@H]1CC[C@@]2([H])[C@]3([H])C[C@H](C)C4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C[C@]12C)=O)=O
Molecular Weight (g/mol) 344.49
Formula Weight 344.49
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.