missing translation for 'onlineSavingsMsg'
Learn More

CHEMGENE MedLab Multi-Surface Disinfectant Wipes

Product Code. 30245735 Shop All Chemgene Products
Change view
Click to view available options
Dimensions (L x W):
298 x 141 mm
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
30245735 298 x 141 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245735 Supplier CHEMGENE Supplier No. SKU101A
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHEMGENE MedLab Multi-Surface Disinfectant Wipes provide fast-acting efficacy against a range of pathogens in 60 seconds, decontaminating surfaces and equipment and denaturing DNA/RNA.

  • Next-generation CHEMGENE HLD4L and HLD4H, CHEMGENE MedLab is a range of surface disinfectants created specifically to meet the demands of laboratory environments.
  • CHEMGENE MedLab Multi-Surface Disinfectant Wipes are proven to be effective under the following microbiological test conditions; Bactericidal: EN 13727, EN 16615. Legionella: EN 13623. Yeasticidal: EN 13624, EN 16615. Fungicidal: EN 13624. Virucidal against all enveloped viruses: EN 14476, EN 16615.
  • Fast-acting efficacy in 60 seconds
  • Denatures DNA/RNA
  • Biodegradable formulation and plastic-free plant-based wipe substrate
  • Large absorbent wipes disinfect 1m² per wipe
  • Manufactured in the UK
  • Non-corrosive
  • Alcohol-free
  • No rinsing required
  • Compatible with a wide range of materials including mild steel, stainless steel, aluminium, copper, nylon, acetal and rubber
  • End-user resources and protocols available to promote correct use and compliance
  • Compliant with REACH, CLP and biocidal regulations in the UK and EU

Specifications

Product Type Disinfectant Wipe
Length (English) 11.7 in.
Length (Metric) 298 mm
Width (English) 7.2 in.
Width (Metric) 185 mm
Material Viscose wipe fabric
Autoclavable Autoclavable
Basis Weight 45 gsm
Container Type Flow wrap
Content And Storage Reseal pack after use to avoid drying out.
Disposable Reusable
Dimensions (L x W) 298 x 141 mm
Format Disinfectant wipes (plastic-free)
For Use With (Application) Cleaning and disinfection of hard surfaces and equipment
Saturation Solution 10% CHEMGENE disinfectant solution
Size 200 x 180 mm
Sterility Non-sterile
Green Features Biodegradable wipe and liquid, plastic-free, made in the UK
pH Range 10.0 to 11.0
Show More Show Less
Product Identifier
  • CHEMGENE MedLab Surface Disinfectant Wipes
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Alkyl (C12-16) dimethylbenzyl ammonium chloride, Didecyldimethylammonium chloride, N-(3-Aminopropyl)

Use biocides safely. Always read the product label and information before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.